Idex® Plugs for 1/4-28 Flat-Bottom Ports with Specialty Heads
Available in various head styles to suit your application
  • Choose from ETFE or PEEK construction
Use the plugs to close off unused ports in valves and multiport connectors.
850.00 - ₹4,600.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Material Description
Availability Pricing
Idex Port Plug, 5/16" Hex Head, Natural ETFE, 1/4-28 Flat-Bottom; 1/EA 0201833 Mfr # P-123
COMPARE
Natural ETFE Port Plug, 5/16" Hex Head, Natural ETFE, 1/4-28 Flat-Bottom; 1/EA
₹850.00INR / Each
Idex Port Plug, FlushNut™ Head, Natural PEEK, 1/4-28 Flat-Bottom; 1/EA 0201836 Mfr # P-321
COMPARE
Natural PEEK Port Plug, FlushNut™ Head, Natural PEEK, 1/4-28 Flat-Bottom; 1/EA
₹4,600.00INR / Each