Idex Port Plug, FlushNut™ Head, Natural PEEK, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Port Plug, FlushNut™ Head, Natural PEEK, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-321| Item: 0201836
Available in various head styles to suit your application
₹4,600.00INR / Each
₹4,600.00INR / Each
Product Description
Key Features
  • Choose from ETFE or PEEK construction
  • Flushnut™ head
  • PEEK construction
  • 1 year warranty
More About this Item
Use the plugs to close off unused ports in valves and multiport connectors.
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorNatural
  • Compatible With1/4-28 flat-bottom port
  • DescriptionPort Plug, FlushNut™ Head, Natural PEEK, 1/4-28 Flat-Bottom; 1/EA