Idex® PEEK Bottom-of-the-Bottle™ Solvent Filters
Protect your system from damaging particulates
  • Filters are biocompatible—constructed of 100% PEEK polymer
Filters allow solvents to be drawn within 0.080” (2.0mm) of the bottom of your solvent bottle. Use the 2 µm frit on the side for a 1/8” OD helium sparging line. Filters have 30 mL/min max flow rate.
5,500.00 - ₹8,300.00INR / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Connections Pore Size (micron)
Availability Pricing
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, PEEK, 2 µm, 1/8" OD Tubing; 1/EA WW-42711-40 Mfr # A-435
COMPARE
1/8\" OD tubing 2
₹6,000.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, PEEK, 2 µm, 3/16" OD Tubing; 1/EA WW-42711-41 Mfr # A-436
COMPARE
3/16\" OD tubing 2
₹6,000.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, Small Neck, PEEK, 2 µm, 1/8" OD Tubing; 1/EA WW-42711-42 Mfr # A-437
COMPARE
1/8\" OD tubing 2
₹5,800.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, Small Neck, PEEK, 10 µm, 1/8" OD Tubing; 1/EA WW-42711-43 Mfr # A-438
COMPARE
1/8\" OD tubing 10
₹6,000.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, PEEK, 10 µm, 1/8" OD Tubing; 1/EA WW-42711-44 Mfr # A-440
COMPARE
1/8\" OD Tubing 10
₹6,000.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, PEEK, 10 µm, 3/16" OD Tubing; 1/EA WW-42711-45 Mfr # A-441
COMPARE
3/16\" OD tubing 10
₹6,000.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, PEEK, 2 µm, 1/16" OD Tubing; 1/EA WW-42711-48 Mfr # A-450
COMPARE
1/16\" OD tubing 2
₹8,300.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, PEEK, 10 µm, 1/16" OD Tubing; 1/EA WW-42711-49 Mfr # A-451
COMPARE
1/16\" OD tubing 10
₹5,600.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, Small Neck, PEEK, 2 µm, 1/16" OD Tubing; 1/EA WW-42711-50 Mfr # A-452
COMPARE
1/16\" OD tubing 2
₹8,300.00INR / Each
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, Small Neck, PEEK, 10 µm, 1/16" OD Tubing; 1/EA WW-42711-51 Mfr # A-453
COMPARE
1/16\" OD tubing 10
₹5,500.00INR / Each
Showing 1- 10 of 10