Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, Small Neck, PEEK, 10 µm, 1/8" OD Tubing; 1/EA
Representative image only.
Idex Bottom-of-the-Bottle™ Biocompatible Filter Assembly, Small Neck, PEEK, 10 µm, 1/8" OD Tubing; 1/EA
Idex | Mfr: A-438| Item: 4271143
Protect your system from damaging particulates
₹10,700.00INR / Each
₹10,700.00INR / Each
Product Description
Key Features
  • Filters are biocompatible—constructed of 100% PEEK polymer
  • For GL-38 small-neck bottles
  • Use with 1/8" OD tubing
  • 1 year warranty
More About this Item
Filters allow solvents to be drawn within 0.080” (2.0mm) of the bottom of your solvent bottle. Use the 2 µm frit on the side for a 1/8” OD helium sparging line. Filters have 30 mL/min max flow rate.
1 Year Warranty
Specifications
  • Membrane MaterialPEEK
  • Pore Size (micron)10
  • Housing MaterialPEEK
  • Connections1/8" OD tubing
  • Typical Water Flow Rate30 mL/min (max)
  • DescriptionBottom-of-the-Bottle™ Biocompatible Filter Assembly, Small Neck, PEEK, 10 µm, 1/8" OD Tubing; 1/EA