Idex® One-Piece MicroTight® Fittings with Standard Knurl Head
Fingertight coned fittings can withstand up to 9000 psi!
  • Easily tighten fittings without tools
  • Variety of fittings to suit your needs
  • One-piece fittings of a nut and ferrule saves time
MicroTight® One-Piece Fittings
Use MicroTight fittings to make superior fingertight connections with capillary tubing. These fittings are designed to work with 360 µm and 1/32" OD tubing or MicroTight tubing sleeves. MicroTight fittings withstand temperatures up to 125°C (257°F).
2,700.00 - ₹26,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Compatible With Description
Availability Pricing
Idex MicroTight® Fitting, Standard Knurl, Natural PEEK, Tubing Sleeves, 6-32 Coned; 1/EA 0201884 Mfr # F-125
COMPARE
Natural 6-32 coned, for tubing sleeves MicroTight® Fitting, Standard Knurl, Natural PEEK, Tubing Sleeves, 6-32 Coned; 1/EA
₹2,700.00INR / Each
Idex MicroTight® Fitting, Standard Knurl, Red PEEK, 1/32" OD Tubing, 6-32 Coned; 1/EA 0201889 Mfr # F-126S
COMPARE
Red 6-32 coned, for 1/32" OD tubing MicroTight® Fitting, Standard Knurl, Red PEEK, 1/32" OD Tubing, 6-32 Coned; 1/EA
₹2,700.00INR / Each
Idex MicroTight® Fitting, Standard Knurl, Red PEEK, 1/32" OD Tubing, 6-32 Coned; 10/PK 0201890 Mfr # F-126SX
COMPARE
Red 6-32 coned, for 1/32" OD tubing MicroTight® Fitting, Standard Knurl, Red PEEK, 1/32" OD Tubing, 6-32 Coned; 10/PK
₹26,700.00INR / Pkg of 10