Idex MicroTight® Fitting, Standard Knurl, Red PEEK, 1/32" OD Tubing, 6-32 Coned; 10/PK
Representative image only.
Idex MicroTight® Fitting, Standard Knurl, Red PEEK, 1/32" OD Tubing, 6-32 Coned; 10/PK
Idex | Mfr: F-126SX| Item: 0201890
Fingertight coned fittings can withstand up to 9000 psi!
₹26,700.00INR / Pkg of 10
₹26,700.00INR / Pkg of 10
Product Description
Key Features
  • Easily tighten fittings without tools
  • Variety of fittings to suit your needs
  • One-piece fittings of a nut and ferrule saves time
  • For 1/32 OD tubing
  • Red PEEK
  • 10/PK
  • 1 year warranty
More About this Item
MicroTight® One-Piece Fittings
Use MicroTight fittings to make superior fingertight connections with capillary tubing. These fittings are designed to work with 360 µm and 1/32" OD tubing or MicroTight tubing sleeves. MicroTight fittings withstand temperatures up to 125°C (257°F).
1 Year Warranty
Specifications
  • MaterialPEEK
  • ColorRed
  • Compatible With6-32 coned, for 1/32" OD tubing
  • DescriptionMicroTight® Fitting, Standard Knurl, Red PEEK, 1/32" OD Tubing, 6-32 Coned; 10/PK