IDEX Natural PFA Chromatography Tubing
Constructed with genuine PFA resin
  • Low gas permeability and outstanding resistance to solvents
  • Maximum operating temperature 80ºC
PFA (perfluoroalkoxyalkane) tubing has virtually identical solvent resistance as FEP and PTFE but offers an even higher continuous service temperature (recommended max operating temperature 80ºC), smoother surface texture, and superior polymer purity.
4,100.00 - ₹450,000.00INR / Each
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Length (ft) Tubing ID (mm) Tubing OD (in)
Availability Pricing
Idex Chromatography Tubing, Natural PFA, 1/16" OD x 0.020" ID x 5 ft L WW-02012-59 Mfr # 1500
COMPARE
5 0.5 1/16
₹4,500.00INR / Each
Idex Chromatography Tubing, Natural PFA, 1/16" OD x 0.030" ID x 5 ft L WW-02012-60 Mfr # 1502
COMPARE
5 0.762 1/16
₹4,500.00INR / Each
Idex Chromatography Tubing, Natural PFA, 1/16" OD x 0.040" ID x 5 ft L WW-02012-61 Mfr # 1503
COMPARE
5 1.016 1/16
₹4,100.00INR / Each
Idex 1507L Chromatography Tubing, Natural PFA, 1/16" OD x 0.040" ID x 50 ft L; 1/EA WW-02012-62 Mfr # 1507L
COMPARE
50 1.016 1/16
₹24,000.00INR / Each
Idex 1509-5 Chromatography Tubing, Natural PFA, 1/8" OD x 0.062" ID x 5 ft L; 1/EA WW-02012-64 Mfr # 1509-5
COMPARE
5 1.575 1/8
₹5,800.00INR / Each
Idex 1509L Chromatography Tubing, Natural PFA, 1/8" OD x 0.062" ID x 50 ft L; 1/EA WW-02012-65 Mfr # 1509L
COMPARE
50 1.575 1/8
₹40,000.00INR / Each
Idex 1512L Chromatography Tubing, Natural PFA, 1/16" OD x 0.020" ID x 50 ft L; 1/EA WW-02012-66 Mfr # 1512L
COMPARE
50 0.5 1/16
₹24,000.00INR / Each
Idex 1514L Chromatography Tubing, Natural PFA, 1/16" OD x .030" ID x 50 ft L; 1/EA WW-02012-67 Mfr # 1514L
COMPARE
50 0.762 1/16
₹24,000.00INR / Each
Idex 1514M Chromatography Tubing, Natural PFA, 1/16" OD x 0.030" ID x 1000 ft L; 1/EA WW-02012-68 Mfr # 1514M
COMPARE
1000 0.762 1/16
₹450,000.00INR / Each
Showing 1- 9 of 9