IDEX® High-Purity PFA Tubing
Manufactured from premium-grade PFA
  • Low gas permeability and outstanding resistance to solvents
  • Maximum operating temperature 80ºC
High-Purity PFA tubing, manufactured from premium-grade PFA, is one of the most contaminant-free polymers available. It offers high continuous service temperature (recommended max operating temperature 80ºC), smooth surface texture, and excellent polymer purity.
8,400.00 - ₹985,000.00INR / Each
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (mm) Tubing OD (in)
Availability Pricing
Idex 1622-5 Chromatography Tubing, High Purity PFA, 1/16" OD x 0.020" ID x 5 ft L; 1/EA 0201269 Mfr # 1622-5
COMPARE
5 0.5 1/16
₹8,400.00INR / Each
Idex 1622L Chromatography Tubing, High Purity PFA, 1/16" OD x 0.020" ID x 50 ft L; 1/EA 0201270 Mfr # 1622L
COMPARE
50 0.5 1/16
₹51,700.00INR / Each
Idex 1632-5 Chromatography Tubing, High Purity PFA, 1/16" OD x 0.030" ID x 5 ft L; 1/EA 0201271 Mfr # 1632-5
COMPARE
5 0.762 1/16
₹8,400.00INR / Each
Idex 1632L Chromatography Tubing, High Purity PFA, 1/16" OD x 0.030" ID x 50 ft L; 1/EA 0201272 Mfr # 1632L
COMPARE
50 0.762 1/16
₹51,700.00INR / Each
Idex 1641-5 Chromatography Tubing, High Purity PFA, 1/8" OD x 0.062" ID x 5 ft L; 1/EA 0201273 Mfr # 1641-5
COMPARE
5 1.575 1/8
₹13,200.00INR / Each
Idex 1641L Chromatography Tubing, High Purity PFA, 1/8" OD x 0.062" ID x 50 ft L; 1/EA 0201274 Mfr # 1641L
COMPARE
50 1.575 1/8
₹80,000.00INR / Each
Idex 1642L Chromatography Tubing, High Purity PFA, 3/16" OD x 0.125" ID x 50 ft L; 1/EA 0201275 Mfr # 1642L
COMPARE
50 3.175 3/16
₹90,100.00INR / Each
Idex 1642XL Chromatography Tubing, High Purity PFA, 3/16" OD x 0.125" ID x 100 ft L; 1/EA 0201276 Mfr # 1642XL
COMPARE
100 3.175 3/16
₹152,000.00INR / Each
Idex 1645L Chromatography Tubing, High Purity PFA, 1/4" OD x 0.188" ID x 50 ft L; 1/EA 0201277 Mfr # 1645L
COMPARE
50 4.775 1/4
₹114,000.00INR / Each
Idex 1645XL Chromatography Tubing, High Purity PFA, 1/4" OD x 0.188" ID x 100 ft L; 1/EA 0201278 Mfr # 1645XL
COMPARE
100 4.775 1/4
₹193,000.00INR / Each
Idex 1632M Chromatography Tubing, High Purity PFA, 1/16" OD x .030" ID x 1000 ft L; 1/EA 0201741 Mfr # 1632M
COMPARE
1000 0.762 1/16
₹985,000.00INR / Each
Showing 1- 11 of 11