Idex® High-Pressure UNF(F) to UNF Adapters with Fitting
Maintain secure connections even under high pressure
  • Securely join together fluid pathway components
  • Manufactured from inert polymers
  • Holds tight connection even through vibration
Made from PEEK, these adapters are durable and have excellent chemical resistance.
9,600.00 - ₹20,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Thread size UNF(F) (port 1) (InchShort) Thread size UNF(F) (port 2) (InchShort) Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Threaded Adapter with Fitting, PEEK, 0.020" ID, 10-32 Coned (F) to 1/4-28 Flat Bottom (F) 0201402 Mfr # P-627
COMPARE
10-32 Coned 1/4-28 Flat Bottom
₹9,600.00INR / Each
Idex Threaded Adapter with Fitting, PEEK, 0.040" ID, 10-32 Coned (F) to 1/4-28 Flat Bottom (M) 0202440 Mfr # P-669
COMPARE
10-32 Coned 1/4-28 Flat Bottom
₹10,000.00INR / Each
Idex Threaded Adapter with Fitting, PEEK, Tapered, 5/16-24 Flat Bottom (F) to 1/2-20 Flat Bottom (F) 0202505 Mfr # U-659
COMPARE
5/16-24 Flat Bottom 1/2-20 Flat Bottom
₹20,700.00INR / Each