Idex Threaded Adapter with Fitting, PEEK, Tapered, 5/16-24 Flat Bottom (F) to 1/2-20 Flat Bottom (F)
Representative image only.
Idex Threaded Adapter with Fitting, PEEK, Tapered, 5/16-24 Flat Bottom (F) to 1/2-20 Flat Bottom (F)
Idex | Mfr: U-659| Item: 0202505
Maintain secure connections even under high pressure
₹20,700.00INR / Each
₹20,700.00INR / Each
Product Description
Includes
One fitting 02021-03
Key Features
  • Securely join together fluid pathway components
  • Manufactured from inert polymers
  • Holds tight connection even through vibration
  • Included fitting accepts 1/4" OD tubing
  • Thru hole tapers from 0.188 to 0.125" (4.8 to 3.2 mm)
  • 42 µL swept volume
  • 1 year warranty
More About this Item
Made from PEEK, these adapters are durable and have excellent chemical resistance.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 1) (InchShort)5/16-24 Flat Bottom
  • Thread size UNF(F) (port 2) (InchShort)1/2-20 Flat Bottom
  • Compression Size / Tube Od (Port 1) (in)1/4
  • ColorNatural
  • Max Operating Pressure (PSI)250
  • DescriptionThreaded Adapter with Fitting, PEEK, Tapered, 5/16-24 Flat Bottom (F) to 1/2-20 Flat Bottom (F)