Idex® High-Pressure True ZDV MicroTight® Adapter Bodies with 6-32 Coned Configuration
Create a high-pressure connection between 1/16" OD tubing and capillary tubing
  • Securely join together fluid pathway components
  • Manufactured from PEEK
  • Holds tight connection even through vibration
Port 1 is 10-32 coned for 1/16" OD tubing.
9,000.00 - ₹9,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color
Availability Pricing
Idex MicroTight Adapter Body, True ZDV, Red PEEK, 1/16 to 1/32" OD, 10-32 to 6-32 0202469 Mfr # P-881-01
COMPARE
Red
₹9,000.00INR / Each
Idex MicroTight Adapter Body, True ZDV, Blue PEEK, 1/16" to 360 µm OD, 10-32 to 6-32 0202470 Mfr # P-882-01
COMPARE
Blue
₹9,200.00INR / Each