Idex MicroTight Adapter Body, True ZDV, Red PEEK, 1/16 to 1/32" OD, 10-32 to 6-32
Representative image only.
Idex MicroTight Adapter Body, True ZDV, Red PEEK, 1/16 to 1/32" OD, 10-32 to 6-32
Idex | Mfr: P-881-01| Item: 0202469
Create a high-pressure connection between 1/16" OD tubing and capillary tubing
₹9,000.00INR / Each
₹9,000.00INR / Each
Product Description
Key Features
  • Securely join together fluid pathway components
  • Manufactured from PEEK
  • Holds tight connection even through vibration
  • Max pressure 5000 psi (345 bar)
  • 1 year warranty
More About this Item
Port 1 is 10-32 coned for 1/16" OD tubing.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 2) (InchShort)6-32 Coned
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Volume (µL)0
  • ColorRed
  • Max Operating Pressure (PSI)5000
  • DescriptionMicroTight Adapter Body, True ZDV, Red PEEK, 1/16 to 1/32" OD Tubing, 10-32 Coned to 6-32 Coned; 1/EA