Idex® High-Pressure NanoTight® Union
Unique design greatly reduces tubing misalignment for greater connection
  • Connect NanoTight tubing sleeves or 1/16" OD tubing
The webbed thru-hole minimizes breaking of fused silica tubing resulting in fewer blockages and fewer back pressure issues.
15,300.00 - ₹18,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Idex High-Pressure Union Assembly, for NanoTight Sleeves or 1/16" OD Tubing, PEEK, 0.005" ID, 10-32 Coned; 1/EA 0201447 Mfr # P-779
COMPARE
High-Pressure Union Assembly, for NanoTight Sleeves or 1/16" OD Tubing, PEEK, 0.005" ID, 10-32 Coned; 1/EA
₹18,500.00INR / Each
Idex High-Pressure Union Body, for NanoTight Sleeves or 1/16" OD Tubing, PEEK, 0.005" ID, 10-32 Coned; 1/EA 0202465 Mfr # P-779-01
COMPARE
High-Pressure Union Body, for NanoTight Sleeves or 1/16" OD Tubing, PEEK, 0.005" ID, 10-32 Coned; 1/EA
₹15,300.00INR / Each