Idex High-Pressure Union Assembly, for NanoTight Sleeves or 1/16" OD Tubing, PEEK, 0.005" ID, 10-32 Coned; 1/EA
Representative image only.
Idex High-Pressure Union Assembly, for NanoTight Sleeves or 1/16" OD Tubing, PEEK, 0.005" ID, 10-32 Coned; 1/EA
Idex | Mfr: P-779| Item: 0201447
Unique design greatly reduces tubing misalignment for greater connection
₹18,500.00INR / Each
₹18,500.00INR / Each
Product Description
Includes
Two fitting assemblies F-331N.
Key Features
  • Connect NanoTight tubing sleeves or 1/16" OD tubing
  • Frit volume 6.1 µL
  • 1 month warranty
  • 1 month warranty
More About this Item
The webbed thru-hole minimizes breaking of fused silica tubing resulting in fewer blockages and fewer back pressure issues.
1 Month Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 1) (InchShort)10-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorNatural
  • Max Operating Pressure (PSI)5000
  • Orifice Dimensions (mm)0.125
  • DescriptionHigh-Pressure Union Assembly, for NanoTight Sleeves or 1/16" OD Tubing, PEEK, 0.005" ID, 10-32 Coned; 1/EA