Idex® High-Pressure MicroTight® Unions for MicroTight Sleeves
Secure connect capillary tubing even under high pressure
  • Holds tight connection even through vibration
  • Withstands up to 4000 psi (276 bar)
These Idex® high-pressure MicroTight® unions for MicroTight sleeves provide a tight, secure connection in your chromatography fluid path.
6,400.00 - ₹7,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Connection Type Max Pressure (PSI) Tubing OD (in)
Availability Pricing
Idex High-Pressure MicroTight Union Assembly, True ZDV for MicroTight Sleeves, PEEK, 6-32 Coned; 1/EA WW-02014-33 Mfr # P-720
COMPARE
HPLC/Chromatography
₹7,700.00INR / Each
Idex High-Pressure MicroTight Union Body, True ZDV for MicroTight Sleeves, PEEK, 6-32 Coned; 1/EA WW-02024-51 Mfr # P-720-01
COMPARE
HPLC/Chromatography
₹6,400.00INR / Each