Idex High-Pressure MicroTight Union Body, True ZDV for MicroTight Sleeves, PEEK, 6-32 Coned; 1/EA
Representative image only.
Idex High-Pressure MicroTight Union Body, True ZDV for MicroTight Sleeves, PEEK, 6-32 Coned; 1/EA
Idex | Mfr: P-720-01| Item: 0202451
Secure connect capillary tubing even under high pressure
₹11,600.00INR / Each
₹11,600.00INR / Each
Product Description
Key Features
  • Holds tight connection even through vibration
  • Withstands up to 4000 psi (276 bar)
  • Union body with 6-32 coned configuration for MicroTight sleeves
  • 1 year warranty
More About this Item
These Idex® high-pressure MicroTight® unions for MicroTight sleeves provide a tight, secure connection in your chromatography fluid path.
Minimum order quantity 5.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Number Of Ports2
  • Thread size UNF(F) (port 1) (InchShort)6-32 Coned
  • Compression Size / Tube Od (Port 1) (in)1/500
  • ColorNatural
  • Max Operating Pressure (PSI)4000
  • DescriptionIdex P-720-01 High-Pressure MicroTight Union Body, True ZDV for MicroTight Sleeves, PEEK, 6-32 Coned; 1/EA