Idex® Flangeless Standard Knurl Male Nuts, 5/16-24 Flat-Bottom Connection
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1,800.00 - ₹1,900.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Fitting Material
Availability Pricing
Idex Flangeless Male Nut, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom; 1/EA 0201924 Mfr # P-130
COMPARE
Natural 1/8 PEEK
₹1,900.00INR / Each
Idex Flangeless Male Nut, Standard Knurl, Natural PEEK, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 1/Ea 0201925 Mfr # P-132
COMPARE
Natural 3/16 PEEK
₹1,800.00INR / Each
Idex Flangeless Male Nut, Standard Knurl, Natural PEEK, 1/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA 0201929 Mfr # P-136
COMPARE
Natural 1/16 PEEK
₹1,900.00INR / Each
Idex Flangeless Male Nut, Standard Knurl, Natural Acetal, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA 1202982 Mfr # P-138
COMPARE
Natural 3/16 Acetal
₹1,800.00INR / Each