Idex Flangeless Male Nut, Standard Knurl, Natural Acetal, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Standard Knurl, Natural Acetal, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA
Idex | Mfr: P-138| Item: 1202982
Eliminate the need to flange tubing
₹1,800.00INR / Each
₹1,800.00INR / Each
Product Description
Key Features
  • Removable and reusable—saving you money!
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)5/16-24 Flat-Bottom
  • Compression Size / Tube Od (Port 1) (in)3/16
  • ColorNatural
  • DescriptionFlangeless Male Nut, Standard Knurl, Natural Acetal, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA