Idex® Flangeless Standard Knurl Fitting Assemblies, 5/16-24 Flat-Bottom Connection
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
2,400.00 - ₹22,800.00INR / Pkg of 10
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Connection Type Description
Availability Pricing
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 10/PK 0201494 Mfr # XP-132X
COMPARE
3/16 HPLC/Chromatography Flangeless Fitting, Standard Knurl, Natural PEEK, 4 mm OD Tubing, 5/16-24 Flat-Bottom
₹22,400.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom; 1/EA 0202057 Mfr # XP-130
COMPARE
1/8 HPLC/Chromatography Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom
₹2,400.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom; 10/PK 0202058 Mfr # XP-130X
COMPARE
1/8 HPLC/Chromatography Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom
₹22,800.00INR / Pkg of 10
Idex Flangeless Fitting, Standard Knurl, Natural PEEK Nut with Blue ETFE Ferrule for 3/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA 0202059 Mfr # XP-132
COMPARE
3/16 HPLC/Chromatography Flangeless Fitting, Standard Knurl, Natural PEEK Nut with Blue ETFE Ferrule for 3/16" OD Tubing, 5/16-24 Flat-Bottom
₹2,400.00INR / Each
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/16" OD Tubing, 5/16-24 Flat-Bottom; 10/PK 0202061 Mfr # XP-136X
COMPARE
1/16 HPLC/Chromatography Flangeless Fitting, Standard Knurl, Natural PEEK, 1/16" OD Tubing, 5/16-24 Flat-Bottom
₹22,800.00INR / Pkg of 10