Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom; 1/EA
Idex | Mfr: XP-130| Item: 0202057
Eliminate the need to flange tubing
₹2,400.00INR / Each
₹2,400.00INR / Each
Buy It Together
Current Item
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom; 1/EA
Idex Flangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom; 1/EA
0202057
₹2,400.00INR / Each
+ + =
Total Price 5270.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Yellow ETFE ferrule 02019-82.
Key Features
  • Removable and reusable—saving you money!
  • Natural PEEK
  • For 1/8" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)5/16-24 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8
  • ColorNatural
  • DescriptionFlangeless Fitting, Standard Knurl, Natural PEEK, 1/8" OD Tubing, 5/16-24 Flat-Bottom