Idex® Flangeless Short Head Male Nuts, M6 Flat-Bottom Connection for 1/16" OD Tubing
Short head is ideal for tight-space applications
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
510.00 - ₹11,400.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Description
Availability Pricing
Idex Flangeless Male Nut, Short Head, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA 0201944 Mfr # P-207S
COMPARE
Acetal
₹510.00INR / Each
Idex Flangeless Male Nut, Short Head, Black Acetal, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK 0201945 Mfr # P-207SX
COMPARE
Acetal
₹4,800.00INR / Pkg of 10
Idex Flangeless Male Nut, Short Head, Black PEEK, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA 0201978 Mfr # P-247
COMPARE
PEEK
₹1,300.00INR / Each
Idex Flangeless Male Nut, Short Head, Black PEEK, 1/16" OD Tubing, M6 Flat-Bottom; 10/PK 0201979 Mfr # P-247X
COMPARE
PEEK
₹11,400.00INR / Pkg of 10