Idex Flangeless Male Nut, Short Head, Black PEEK, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Male Nut, Short Head, Black PEEK, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Idex | Mfr: P-247| Item: 0201978
Short head is ideal for tight-space applications
₹1,300.00INR / Each
₹1,300.00INR / Each
Buy It Together
Current Item
Idex Flangeless Male Nut, Short Head, Black PEEK, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
Idex Flangeless Male Nut, Short Head, Black PEEK, 1/16" OD Tubing, M6 Flat-Bottom; 1/EA
0201978
₹1,300.00INR / Each
+ =
Total Price 1790.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
  • Black PEEK
  • 1/EA
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Connection TypeHPLC/Chromatography
  • Compression Size / Tube Od (Port 1) (in)1/16
  • Thread Size ISO Metric (M) (Port 2) (mm)6
  • ColorBlack