Idex® Flangeless Short Head Fitting Assemblies, 1/4-28 Flat-Bottom Connection for 1/16" OD Tubing
Eliminate the need to flange tubing
  • Removable and reusable—saving you money!
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1,300.00 - ₹16,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Fitting Material Description
Availability Pricing
Idex Flangeless Fitting, Short Head, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0201497 Mfr # XP-235X
COMPARE
Natural PEEK
₹16,800.00INR / Pkg of 10
Idex Flangeless Fitting, Short Head, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/Each 0202070 Mfr # XP-208
COMPARE
Black Acetal
₹1,300.00INR / Each
Idex Flangeless Fitting, Short Head, Natural PEEK, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202078 Mfr # XP-235
COMPARE
Natural PEEK
₹1,700.00INR / Each