Idex Flangeless Fitting, Short Head, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/Each
Representative image only.
Idex Flangeless Fitting, Short Head, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/Each
Idex | Mfr: XP-208| Item: 0202070
Eliminate the need to flange tubing
₹1,300.00INR / Each
₹1,300.00INR / Each
Buy It Together
Current Item
Idex Flangeless Fitting, Short Head, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/Each
Idex Flangeless Fitting, Short Head, Black Acetal, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/Each
0202070
₹1,300.00INR / Each
+ + + =
Total Price 17350.00 ADD 4 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Blue ETFE ferrules 02019-33.
Key Features
  • Removable and reusable—saving you money!
  • Black acetal
  • 1 Each
  • 1 year warranty
More About this Item
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components.
1 Year Warranty
Specifications
  • Fitting MaterialAcetal
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/16
  • ColorBlack