Idex® Flangeless Ferrules, 5/16-24 Flat-Bottom Connection for 3/16" OD Tubing
Use with male nuts for a leak-tight connection.
  • Excellent for rigid and semi-rigid tubing
Flangeless ferrules are most commonly used with large bore fittings with a flat-bottom port. It is the most frequently used threaded port configuration for low pressure connections.
Use with male nuts for a leak-tight connection.
510.00 - ₹4,800.00INR / Pkg of 10
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Flangeless Ferrule, Blue ETFE, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 10/PK 0201366 Mfr # P-133X
COMPARE
Blue 3/16 5/16-24 flat bottom
₹4,600.00INR / Pkg of 10
Idex Flangeless Ferrule, Natural ETFE, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 1/EA 0201927 Mfr # P-133N
COMPARE
Natural 3/16 5/16-24 flat bottom
₹510.00INR / Each
Idex Flangeless Ferrule, Natural ETFE, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 10/PK 0201928 Mfr # P-133NX
COMPARE
Natural 3/16 5/16-24 flat bottom
₹4,800.00INR / Pkg of 10