Idex Flangeless Ferrule, Natural ETFE, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 10/PK
Representative image only.
Idex Flangeless Ferrule, Natural ETFE, 3/16" OD Tubing, 5/16-24 Flat-Bottom; 10/PK
Idex | Mfr: P-133NX| Item: 0201928
Use with male nuts for a leak-tight connection.
₹4,800.00INR / Pkg of 10
₹4,800.00INR / Pkg of 10
Product Description
Key Features
  • Excellent for rigid and semi-rigid tubing
  • Natural ETFE
  • 10/PK
  • 1 year warranty
More About this Item
Flangeless ferrules are most commonly used with large bore fittings with a flat-bottom port. It is the most frequently used threaded port configuration for low pressure connections.
Use with male nuts for a leak-tight connection.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)5/16-24 flat bottom
  • Compression Size / Tube Od (Port 1) (in)3/16