Idex® Flangeless Ferrules, 10-32 or 1/4-28 Flat-Bottom Connection, ETFE
No need to flange tubing
  • Excellent for connecting rigid or semi-rigid tubing
Use with male nuts for a leak-tight connection.
490.00 - ₹4,700.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Color Compression Size / Tube Od (Port 1) (in) Thread size UNF(M) (port 2) (InchShort)
Availability Pricing
Idex Flangeless Ferrule, Short, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0201382 Mfr # P-340X
COMPARE
Natural 1/8 1/4-28 flat bottom
₹4,600.00INR / Pkg of 10
Idex Flangeless Ferrule, Natural ETFE, 1/16" OD Tubing, 10-32 or 1/4-28 Flat-Bottom; 1/EA 0201974 Mfr # P-240
COMPARE
Natural 1/16 1/4-28 flat bottom
₹490.00INR / Each
Idex Flangeless Ferrule, Natural ETFE, 1/16" OD Tubing, 10-32 or 1/4-28 Flat-Bottom; 10/PK 0201975 Mfr # P-240X
COMPARE
Natural 1/16 1/4-28 flat bottom
₹4,700.00INR / Pkg of 10
Idex Flangeless Ferrule, Short, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA 0202031 Mfr # P-340
COMPARE
Natural 1/8 1/4-28 flat bottom
₹500.00INR / Each