Idex Flangeless Ferrule, Short, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Representative image only.
Idex Flangeless Ferrule, Short, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Idex | Mfr: P-340| Item: 0202031
No need to flange tubing
₹500.00INR / Each
₹500.00INR / Each
Buy It Together
Current Item
Idex Flangeless Ferrule, Short, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
Idex Flangeless Ferrule, Short, Natural ETFE, 1/8" OD Tubing, 1/4-28 Flat Bottom; 1/EA
0202031
₹500.00INR / Each
+ =
Total Price 1600.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Excellent for connecting rigid or semi-rigid tubing
  • 1/4-28 flat bottom
  • For 1/8" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
Use with male nuts for a leak-tight connection.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/4-28 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/8