Idex® Flangeless Ferrules, 1/2-20 Flat-Bottom Connection, ETFE
Excellent for rigid and semi-rigid tubing
  • Excellent for connecting rigid or semi-rigid tubing
Headless super flangeless nut eliminates loosening of fittings due to tube twisting, holds tight even during vibration, and delivers the highest pressure holding for flat bottom fittings. Use with male nuts for a leak-tight connection.
710.00 - ₹10,700.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Compression Size / Tube Od (Port 1) (in) Fitting Material Description
Availability Pricing
Idex Flangeless Ferrule, Natural ETFE, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA 0202047 Mfr # U-650
COMPARE
1/4 ETFE Flangeless Ferrule, Natural ETFE, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
₹710.00INR / Each
Idex Flangeless Ferrule, Natural ETFE, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 10/PK 0202048 Mfr # U-650X
COMPARE
1/4 ETFE Flangeless Ferrule, Natural ETFE, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 10/PK
₹6,800.00INR / Pkg of 10
Idex Flangeless Ferrule, Natural ETFE, 5/16" OD Tubing, 1/2-20 Flat-Bottom; 10/PK 0202051 Mfr # U-660X
COMPARE
5/16 ETFE Flangeless Ferrule, Natural ETFE, 5/16" OD Tubing, 1/2-20 Flat-Bottom; 10/PK
₹10,700.00INR / Pkg of 10