Idex Flangeless Ferrule, Natural ETFE, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
Representative image only.
Idex Flangeless Ferrule, Natural ETFE, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA
Idex | Mfr: U-650| Item: 0202047
Excellent for rigid and semi-rigid tubing
₹710.00INR / Each
₹710.00INR / Each
Product Description
Key Features
  • Excellent for connecting rigid or semi-rigid tubing
  • Natural ETFE
  • For 1/4" OD tubing
  • 1/EA
  • 1 year warranty
More About this Item
Headless super flangeless nut eliminates loosening of fittings due to tube twisting, holds tight even during vibration, and delivers the highest pressure holding for flat bottom fittings. Use with male nuts for a leak-tight connection.
1 Year Warranty
Specifications
  • Fitting MaterialETFE
  • Connection TypeHPLC/Chromatography
  • Thread size UNF(M) (port 2) (InchShort)1/2-20 flat bottom
  • Compression Size / Tube Od (Port 1) (in)1/4
  • ColorNatural
  • DescriptionFlangeless Ferrule, Natural ETFE, 1/4" OD Tubing, 1/2-20 Flat-Bottom; 1/EA