Idex® Flanged Fitting Accessories
Use with flanged tubing
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
These chromatography fittings of nuts and washers are used with flanged tubing. Flanged nuts are available in Delrin® acetal to resist cross-threading or loosening while in use or choose ethylene tetrafluoroethylene (ETFE) for chemically aggressive environments. The nuts come with stainless steel washers.
290.00 - ₹7,000.00INR / Pkg of 10
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
Idex Flanged Washer, SS, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK 0202553 Mfr # P-1087X
COMPARE
Stainless steel M6 flat-bottom flanged nut for 1/8" OD tubing Flanged Washer, SS, 1/8" OD Tubing, M6 Flat-Bottom; 10/PK
₹7,000.00INR / Pkg of 10
Idex Flanged Washer, SS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202561 Mfr # P-407-01
COMPARE
Stainless steel 1/4-28 flat-bottom flanged nut for 1/16" OD tubing Flanged Washer, SS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
₹420.00INR / Each
Idex Flanged Washer, SS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202562 Mfr # P-407X
COMPARE
Stainless steel 1/4-28 flat-bottom flanged nut for 1/16" OD tubing Flanged Washer, SS, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
₹4,400.00INR / Pkg of 10
Idex Flanged Nut, Standard Knurl, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202568 Mfr # P-482T-01
COMPARE
ETFE Washers 02025-61 or 02025-62 Flanged Nut, Standard Knurl, Natural ETFE, 1/16" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
₹760.00INR / Each
Idex Flanged Washer, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA 0202576 Mfr # P-507-01
COMPARE
Stainless steel 1/4-28 flat-bottom flanged nut for 1/8" OD tubing Flanged Washer, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
₹290.00INR / Each
Idex Flanged Washer, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK 0202577 Mfr # P-507X
COMPARE
Stainless steel 1/4-28 flat-bottom flanged nut for 1/8" OD tubing Flanged Washer, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 10/PK
₹3,400.00INR / Pkg of 10