Idex Flanged Washer, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Representative image only.
Idex Flanged Washer, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Idex | Mfr: P-507-01| Item: 0202576
Use with flanged tubing
₹290.00INR / Each
₹290.00INR / Each
Product Description
Key Features
  • Choice of fittings for 1/16" or 1/8" OD tubing
  • Multiple head styles and materials
  • Choice of colors
  • For 1/8" OD tubing
  • 1/4-28 flat-bottom
  • 1/EA
  • 1 year warranty
More About this Item
These chromatography fittings of nuts and washers are used with flanged tubing. Flanged nuts are available in Delrin® acetal to resist cross-threading or loosening while in use or choose ethylene tetrafluoroethylene (ETFE) for chemically aggressive environments. The nuts come with stainless steel washers.
1 Year Warranty
Specifications
  • MaterialStainless steel
  • Compatible With1/4-28 flat-bottom flanged nut for 1/8" OD tubing
  • DescriptionFlanged Washer, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA