Idex® Extender Tools for Fittings
Easily reach hard-to-reach places to tighten fitting nuts
  • Choose from aluminum or acetal construction
Tighten most of the knurled nuts in hard-to-reach places for leak-tight connections using extender tools. Simply match up the head style to the tool. Use extender tools to torque driver 02022-80 for tightening to a specific amount of tightening torque or as stand-alone units.
1,600.00 - ₹28,400.00INR / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
Idex Extender Tool for Micro Headless Nuts; 1/EA WW-02025-38 Mfr # N-290
COMPARE
Aluminum Micro headless nut knurl head Extender Tool for Micro Headless Nuts; 1/EA
₹5,800.00INR / Each
Idex Extender Tool to Torque Driver for Micro Headless Nuts; 1/EA WW-02025-39 Mfr # N-291
COMPARE
Aluminum Micro headless nut knurl head Extender Tool to Torque Driver for Micro Headless Nuts; 1/EA
₹15,400.00INR / Each
Idex Extender Tool to Torque Driver for 1/4" Hex Nuts; 1/EA WW-02025-41 Mfr # P-268
COMPARE
Aluminum 1/4 hex head Extender Tool to Torque Driver for 1/4\" Hex Nuts; 1/EA
₹28,400.00INR / Each
Idex Extender Tool for Micro Nuts; 1/EA WW-02025-42 Mfr # P-277
COMPARE
Aluminum Micro nut knurl head Extender Tool for Micro Nuts; 1/EA
₹11,500.00INR / Each
Idex Extender Tool to Torque Driver for Female Nuts; 1/EA WW-02025-43 Mfr # P-278
COMPARE
Aluminum Female nut knurl head Extender Tool to Torque Driver for Female Nuts; 1/EA
₹10,600.00INR / Each
Idex Extender Tool to Torque Driver for Micro Nuts; 1/EA WW-02025-44 Mfr # P-279
COMPARE
Aluminum Micro nut knurl head Extender Tool to Torque Driver for Micro Nuts; 1/EA
₹17,400.00INR / Each
Idex Extender Tool to Torque Driver for Standard Nuts; 1/EA WW-02025-45 Mfr # P-291
COMPARE
Aluminum Standard nut knurl head Extender Tool to Torque Driver for Standard Nuts; 1/EA
₹10,500.00INR / Each
Idex Extender Tool to Torque Driver for Headless Nuts; 1/EA WW-02025-46 Mfr # P-292
COMPARE
Aluminum Headless nut head Extender Tool to Torque Driver for Headless Nuts; 1/EA
₹10,900.00INR / Each
Idex Extender Tool for Headless Nuts; 1/EA WW-02025-47 Mfr # P-297
COMPARE
Aluminum Headless nut head Extender Tool for Headless Nuts; 1/EA
₹2,400.00INR / Each
Idex Extender Tool for Standard Head Nuts, Acetal; 1/EA WW-02025-48 Mfr # P-298
COMPARE
Acetal Standard nut knurl head Extender Tool for Standard Head Nuts, Acetal; 1/EA
₹1,600.00INR / Each
Idex Extender Tool for Standard Head Nuts, Aluminum; 1/EA WW-02025-49 Mfr # P-299
COMPARE
Aluminum Standard nut knurl head Extender Tool for Standard Head Nuts, Aluminum; 1/EA
₹3,200.00INR / Each
Idex Extender Tool for 8 mm Hex; 1/EA WW-02025-50 Mfr # P-398
COMPARE
Aluminum 8 mm hex head Extender Tool for 8 mm Hex; 1/EA
₹5,800.00INR / Each
Idex Extender Tool for Standard Head Nuts, Short; 1/EA WW-02025-51 Mfr # P-399
COMPARE
Aluminum Standard nut knurl, short head Extender Tool for Standard Head Nuts, Short; 1/EA
₹3,200.00INR / Each
Showing 1- 13 of 13