Idex Extender Tool to Torque Driver for Micro Nuts; 1/EA
Representative image only.
Idex Extender Tool to Torque Driver for Micro Nuts; 1/EA
Idex | Mfr: P-279| Item: 0202544
Easily reach hard-to-reach places to tighten fitting nuts
₹30,900.00INR / Each
₹30,900.00INR / Each
Product Description
Key Features
  • Choose from aluminum or acetal construction
  • Use with torque driver for micro nuts
  • 1 year warranty
More About this Item
Tighten most of the knurled nuts in hard-to-reach places for leak-tight connections using extender tools. Simply match up the head style to the tool. Use extender tools to torque driver 02022-80 for tightening to a specific amount of tightening torque or as stand-alone units.
1 Year Warranty
Specifications
  • MaterialAluminum
  • Compatible WithMicro nut knurl head
  • DescriptionExtender Tool to Torque Driver for Micro Nuts; 1/EA