IDEX® Ethylene Tetrafluoroethylene (ETFE) Tubing
Withstands the highest pressures of fluoropolymer tubing
  • Excellent chemical resistance
  • Max continuous operating temperature of 176°F (80°C)
ETFE offers low gas permeability, excellent mechanical stability, and high pressure-holding capabilities. Due to its low gas permeability, ETFE is an ideal choice for connecting the system pump to the vacuum degasser which will help prevent regassing of mobile phase solvents in transit.
6,900.00 - ₹654,000.00INR / Each
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Tubing ID (in) Tubing OD (in)
Availability Pricing
Idex 1516XL Chromatography Tubing, Natural ETFE, 1/16" OD x 0.020" ID x 100 ft L; 1/EA 0201295 Mfr # 1516XL
COMPARE
100 1/50 1/16
₹68,900.00INR / Each
Idex 1528XL Chromatography Tubing, Natural ETFE, 1/16" OD x 0.030" ID x 100 ft L; 1/EA 0201299 Mfr # 1528XL
COMPARE
100 1/32 1/16
₹68,900.00INR / Each
Idex Chromatography Tubing, Natural ETFE, 1/8" OD x 0.062" ID x 5 ft L 0201302 Mfr # 1530
COMPARE
5 1/16 1/8
₹10,300.00INR / Each
Idex 1530XL Chromatography Tubing, Natural ETFE, 1/8" OD x 0.062" ID x 100 ft L; 1/EA 0201303 Mfr # 1530XL
COMPARE
100 1/16 1/8
₹106,000.00INR / Each
Idex 1648XL Chromatography Tubing, Natural ETFE, 1/8" OD x 0.093" ID x 100 ft L; 1/EA 0201305 Mfr # 1648XL
COMPARE
100 3/32 1/8
₹106,000.00INR / Each
Idex 1325 Chromatography Tubing Kit, ETFE; 1/EA 0201327 Mfr # 1325
COMPARE
5 1/100 1/16
₹53,600.00INR / Each
Idex 1516 Chromatography Tubing, Natural ETFE, 1/16" OD x 0.020" ID x 5 ft L; 1/EA 0201328 Mfr # 1516
COMPARE
5 1/50 1/16
₹6,900.00INR / Each
Idex 1517 Chromatography Tubing, Natural ETFE, 1/16" OD x 0.040" ID x 5 ft L; 1/EA 0201331 Mfr # 1517
COMPARE
5 1/25 1/16
₹6,900.00INR / Each
Idex 1528 Chromatography Tubing, Natural ETFE, 1/16" OD x 0.030" ID x 5 ft L; 1/EA 0201350 Mfr # 1528
COMPARE
5 1/32 1/16
₹6,900.00INR / Each
Idex 1529 Chromatography Tubing, Natural ETFE, 1/16" OD x 0.010" ID x 5 ft L; 1/EA 0201351 Mfr # 1529
COMPARE
5 1/100 1/16
₹6,900.00INR / Each
Idex 1516L Chromatography Tubing, Natural ETFE, 1/16" OD x 0.020" ID x 50 ft L; 1/EA 0201369 Mfr # 1516L
COMPARE
50 1/50 1/16
₹41,300.00INR / Each
Idex 1517L Chromatography Tubing, Natural ETFE, 1/16" OD x 0.040" ID x 50 ft L; 1/EA 0201375 Mfr # 1517L
COMPARE
50 1/25 1/16
₹41,300.00INR / Each
Idex 1517M Chromatography Tubing, Natural ETFE, 1/16" OD x 0.040" ID x 1000 ft L; 1/EA 0201376 Mfr # 1517M
COMPARE
1000 1/25 1/16
₹654,000.00INR / Each
Idex 1517XL Chromatography Tubing, Natural ETFE, 1/16" OD x 0.040" ID x 100 ft L; 1/EA 0201427 Mfr # 1517XL
COMPARE
100 1/25 1/16
₹70,000.00INR / Each
Idex 1528L Chromatography Tubing, Natural ETFE, 1/16" OD x 0.030" ID x 50 ft L; 1/EA 0201428 Mfr # 1528L
COMPARE
50 1/32 1/16
₹42,100.00INR / Each
Idex 1529L Chromatography Tubing, Natural ETFE, 1/16" OD x 0.010" ID x 50 ft L; 1/EA 0201431 Mfr # 1529L
COMPARE
50 1/100 1/16
₹41,300.00INR / Each
Idex 1529XL Chromatography Tubing, Natural ETFE, 1/16" OD x 0.010" ID x 100 ft L; 1/EA 0201436 Mfr # 1529XL
COMPARE
100 1/100 1/16
₹68,900.00INR / Each
Idex 1530L Chromatography Tubing, Natural ETFE, 1/8" OD x 0.062" ID x 50 ft L; 1/EA 0201471 Mfr # 1530L
COMPARE
50 1/16 1/8
₹63,100.00INR / Each
Idex 1648L Chromatography Tubing, Natural ETFE, 1/8" OD x 0.093" ID x 50 ft L; 1/EA 0201472 Mfr # 1648L
COMPARE
50 3/32 1/8
₹63,100.00INR / Each
Idex Chromatography Tubing, Natural ETFE, 1/8" OD x 0.093" ID x 5 ft L 0201739 Mfr # 1648
COMPARE
5 3/32 1/8
₹10,500.00INR / Each
Showing 1- 20 of 20