Idex® Adapter Assemblies
Connect soft-wall tubing to rigid and semi-rigid tubing
  • Biocompatible flow path with excellent chemical compatibility
  • Soft-wall tubing slides on easily on the narrow adapter body
Barbed adapter body accepts a range of soft-wall tubing.
5,500.00 - ₹12,800.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Barb Size / Tubing Id (Port 2) (mm) Compression Size / Tube Od (Port 1) (in) Orifice Dimensions (mm)
Availability Pricing
Idex Conical Adapter, Natural PEEK/ETFE, 0.020" Bore, 1/16" OD Rigid x 0.020-0.030" ID Soft Tubing; 1/EA 0201454 Mfr # P-794
COMPARE
0.75 1/16 0.5
₹7,900.00INR / Each
Idex Conical Adapter, Natural PEEK/Blue ETFE, 0.040" Bore, 1/16" OD Rigid x 1/16-3/32" ID Soft Tubing; 1/EA 0202382 Mfr # P-797
COMPARE
2.4 1/16 1
₹5,500.00INR / Each
Idex Conical Adapter, Natural PEEK/Blue ETFE, 0.040" Bore, 1/8" OD Rigid x 1/16-3/32" ID Soft Tubing; 1/EA 0202383 Mfr # P-798
COMPARE
2.4 1/8 1
₹5,500.00INR / Each
Idex Conical Adapter, Natural PEEK/Blue ETFE, 0.060" Bore, 1/8" OD Rigid x 3/32-1/8" ID Soft Tubing; 1/EA 0202384 Mfr # P-799
COMPARE
3.2 1/8 1.5
₹5,600.00INR / Each
Idex Adapter Assembly, 1/2-20 Female x 1/4-28 UNF(F) Flat; 1/EA 4271960 Mfr # U-665
COMPARE
₹12,800.00INR / Each