Idex Adapter Assembly, 1/2-20 Female x 1/4-28 UNF(F) Flat; 1/EA

Representative image only.

Idex Adapter Assembly, 1/2-20 Female x 1/4-28 UNF(F) Flat; 1/EA

Idex | Mfr: U-665| Item: 4271960

Connect soft-wall tubing to rigid and semi-rigid tubing

₹12,800.00INR / Each

₹12,800.00INR / Each

Buy It Together

Current Item

Idex Adapter Assembly, 1/2-20 Female x 1/4-28 UNF(F) Flat; 1/EA
Idex Adapter Assembly, 1/2-20 Female x 1/4-28 UNF(F) Flat; 1/EA
4271960

₹12,800.00INR / Each
+ =
Total Price 27400.00 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Biocompatible flow path with excellent chemical compatibility
  • Soft-wall tubing slides on easily on the narrow adapter body
  • Versatile PEEK English adapter assembly to attach connectors with different threads
  • Female 1/2-20 to female 1/4-28 flat-bottom port configuration
  • 1 year warranty

More About this Item

Barbed adapter body accepts a range of soft-wall tubing.

1 Year Warranty
Specifications
  • Fitting MaterialPEEK
  • Flow RateSwept Volume nL 42
  • Max Operating Pressure (PSI)250 (17 bar)
  • DescriptionAdapter Assembly 1/2-20 Female x1/4-28 Female; 1/EA
  • Warranty1 year
Reviews
Q&A