Hydrion Sanitizer Strength Test Stations
Convenient dispenser roll lets you use the exact strip length needed
  • Wall-mounted test centers put sanitizers within easy reach
Federal, State, and Local health regulations require the use of test kits to verify the strength of sanitizer solutions. Hydrion test strips are a quick, reliable, and economocial solution for this. Strips provide a visual color change indication of solution strength. Each 15-ft (4.6-m) roll comes with a color match chart and provides paper for about 100 tests.
16,000.00 - ₹16,200.00INR / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Hydrion CHLORSTART Clorstart Chlorine Sanitizer Test Center 5920058 Mfr # CHLORSTART
COMPARE
Clorstart Chlorine Sanitizer Test Center
₹16,000.00INR / Pkg of 1
Hydrion QUATSTART Quat Sanitizer Testing Center 5920059 Mfr # QUATSTART
COMPARE
Quat Sanitizer Testing Center
₹16,200.00INR / Pkg of 1