Hydrion CHLORSTART Clorstart Chlorine Sanitizer Test Center
Representative image only.
Hydrion CHLORSTART Clorstart Chlorine Sanitizer Test Center
Hydrion | Mfr: CHLORSTART| Item: 5920058
Convenient dispenser roll lets you use the exact strip length needed
₹16,000.00INR / Pkg of 1
₹16,000.00INR / Pkg of 1
Product Description
Key Features
  • Wall-mounted test centers put sanitizers within easy reach
More About this Item
Federal, State, and Local health regulations require the use of test kits to verify the strength of sanitizer solutions. Hydrion test strips are a quick, reliable, and economocial solution for this. Strips provide a visual color change indication of solution strength. Each 15-ft (4.6-m) roll comes with a color match chart and provides paper for about 100 tests.
This starter kit offers everything you need to ensure that your chlorine based sanitizer levels are within code. The storage unit mounts above the sink, keeping the test kits within easy reach. The poster serves as a reminder of proper testing procedures.
Includes wall mounted unit, 3 chlorine test dispensers, mounting hardware and a self adhesive instruction poster.
Specifications
  • TestChlorine, sanitizer
  • Reagent PlatformStrips
  • DescriptionClorstart Chlorine Sanitizer Test Center