Huber TC Immersion Coolers

Reduce tap water consumption
  • Small footprint saves valuable countertop space
  • Ecofriendly-natutal refrigerant and low energy consumption
Huber TC immersion coolers offers a flexible solution for a variety of cooling applications. They are easy to use and ideally suited to quick liquid cooling while offering an environmentally friendly alternative to tap water or dry ice. The coolers are available without a controller for continuous cooling or with controller for greater temperature control.
Compact design combined with carrying handles and flexible hose make these coolers easy to move and reposition as needed. Rugged stainless steel housing and condensing probe will provide years of service.
Call For Price

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cooling Capacity At 0° C (watts) Min Temperature (° C) Power (VAC)
Availability Pricing
Huber TC45E  Immersion Cooler, 115 VAC, 60Hz WW-12751-40 Mfr # 3003.0006.99
COMPARE
240 -45 115
Call For Price
Huber TC45E-F  Immersion Cooler, 115 VAC, 60Hz WW-12751-41 Mfr # 3003.0008.99
COMPARE
240 -45 115
Call For Price
Huber TC50E  Immersion Cooler, 110 VAC, 50/60Hz WW-12751-44 Mfr # 3004.0006.99
COMPARE
300 -50 110
Call For Price
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat