Huber Ministat Cooling/Heating Circulating Baths
Smallest cooling circulator available – perfect for fume hood work
  • Environmentally friendly temperature control units utilize natural refrigerants
  • Combined electronic low-level and user-adjustable overtemperature protection
  • Intuitive Pilot ONE touchscreen controller simplifies tasks
  • Store and load temperature settings to and from a USB stick (not provided)
Function is not compromised for size with the Ministat series circulators. Features of the integrated Pilot ONE controller include a large, 5.7" color, touch-screen display with 11 language support (EN, DE, FR, IT, ES, RU, CN, PT, JP, CZ, and PL), convenient color-coded navigation system, and USB and Ethernet ports for communication and remote control through your tablet, PC, or network.
These baths use environmentally friendly natural refrigerants, air or water depending on the model, while boasting a powerful, variable-speed pressure/suction pump—ideal for external cooling to photometers, viscometers, refractometers, and distillation/extraction apparatus. The bath cover may be removed to heat/cool samples directly in the stainless-steel reservoir, too.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Min Temperature (° C) Description
Availability Pricing
Huber Ministat 125 Refrigerated/Heating Circulating Bath; 115V WW-12125-30 Mfr # 2014.0012.01
COMPARE
150 -25 Refrigerated/Heating Circulating Bath; 115V
Call For Price
Huber Ministat 240 Refrigerated/Heating Circulating Bath; 208V WW-12125-34 Mfr # 2016.0018.01
COMPARE
200 -45 Refrigerated/Heating Circulating Bath; 208V
Call For Price
Huber Ministat 125 Refrigerated/Heating Circulating Bath; 230V WW-12125-37 Mfr # 2014.0011.01
COMPARE
150 -25 Refrigerated/Heating Circulating Bath; 230V