Howard Leight VeriShield™ 100 Series Over-the-Head Earmuffs
Protect your ears in comfort
  • Superior hearing protection in a lightweight design helps lower risk of noise-induced hearing loss
  • Memory foam ear cushions provide superior comfort and protection
  • Adjustable, steel-wire padded headband for durability and comfort
  • Airflow control technology aids to comfort when wearing with hats
  • Large openings of ear cushions are ideal for those with larger ears or hearing aids
These over-the-head noise-reducing earmuffs feature unique memory foam ear cushions that combine denser and softer foam than prior models for greater comfort. Plus, the lightweight cushioned headband with precision micro adjustments is designed to better fit around ears. The earmuffs comfortably fit a wide range of head sizes with about 17% more adjustability than its predecessors.
At the same time, the opening area of the ear cups increased by about 16%, ensuring a good fit for all.
2,700.00 - ₹12,200.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
NRR (dB) Description
Availability Pricing
Howard Leight VS110 VeriShield™ Over-the-Head Earmuff, Low Level/Flat 8632640 Mfr # 1035100-VS
COMPARE
24 VeriShield™ Over-the-Head Earmuff, Low Level/Flat
₹3,200.00INR / Each
Howard Leight VS110F VeriShield™ Over-the-Head Folding Earmuff, Low Level/Flat 8632641 Mfr # 1035102-VS
COMPARE
24 VeriShield™ Over-the-Head Folding Earmuff, Low Level/Flat
₹5,100.00INR / Each
Howard Leight VS120 VeriShield™ Over-the-Head Earmuff, Mid Level 8632642 Mfr # 1035104-VS
COMPARE
26 VeriShield™ Over-the-Head Earmuff, Mid Level
₹2,700.00INR / Each
Howard Leight VS130 VeriShield™ Over-the-Head Earmuff, High Level 8632644 Mfr # 1035108-VS
COMPARE
30 VeriShield™ Over-the-Head Earmuff, High Level
₹10,500.00INR / Each
Howard Leight VS130HV VeriShield™ Over-the-Head High Visibility Earmuff, High Level 8632645 Mfr # 1035110-VS
COMPARE
30 VeriShield™ Over-the-Head High Visibility Earmuff, High Level
₹12,200.00INR / Each