Howard Leight VS130 VeriShield™ Over-the-Head Earmuff, High Level
Representative image only.
Howard Leight VS130 VeriShield™ Over-the-Head Earmuff, High Level
Howard Leight | Mfr: 1035108-VS| Item: 8632644
Protect your ears in comfort
₹10,500.00INR / Each
₹10,500.00INR / Each
Product Description
Key Features
  • Superior hearing protection in a lightweight design helps lower risk of noise-induced hearing loss
  • Memory foam ear cushions provide superior comfort and protection
  • Adjustable, steel-wire padded headband for durability and comfort
  • Airflow control technology aids to comfort when wearing with hats
  • Large openings of ear cushions are ideal for those with larger ears or hearing aids
More About this Item
These over-the-head noise-reducing earmuffs feature unique memory foam ear cushions that combine denser and softer foam than prior models for greater comfort. Plus, the lightweight cushioned headband with precision micro adjustments is designed to better fit around ears. The earmuffs comfortably fit a wide range of head sizes with about 17% more adjustability than its predecessors.
At the same time, the opening area of the ear cups increased by about 16%, ensuring a good fit for all.
Specifications
  • NRR (dB)30
  • DescriptionVeriShield™ Over-the-Head Earmuff, High Level