Honeywell Input and Output Cards for Multiparameter Analyzer
Optimize performance of your Honeywell oxygen meter
  • DO input module for ppm measurement
  • For channel 1 and/or 2
Module slides in dual-input analytical analyzer to take dissolved oxygen measurments.
169,000.00 - ₹175,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Type
Availability Pricing
Honeywell 51453313-501 pH/ORP Input Card 5661102 Mfr # 51453313-501
COMPARE
Input—pH/ORP
₹169,000.00INR / Each
Honeywell 51453319-501 Dissolved Oxygen Input Card, ppm Range 5661106 Mfr # 51453319-501
COMPARE
Input—dissolved oxygen
₹175,000.00INR / Each
Honeywell 51453328-501 Two Relays & 0/4 to 20 mA Output Card, 5661110 Mfr # 51453328-501
COMPARE
Output—two relays & 0/4 to 20 mA
₹171,000.00INR / Each