Honeywell 51453328-501 Two Relays & 0/4 to 20 mA Output Card,
Representative image only.
Honeywell 51453328-501 Two Relays & 0/4 to 20 mA Output Card,
Honeywell | Mfr: 51453328-501| Item: 5661110
Optimize performance of your Honeywell oxygen meter
₹171,000.00INR / Each
₹171,000.00INR / Each
Product Description
Key Features
  • DO input module for ppm measurement
  • For channel 1 and/or 2
  • 1 year warranty
More About this Item
Module slides in dual-input analytical analyzer to take dissolved oxygen measurments.
Optional, additional relay card for the Honeywell Dual-input controller. Output card provides two extra electromechanical relays (comes with two standard), and one 4 to 20/0 to 20 mA output—perfect for connecting to a recorder.
1 Year Warranty
Specifications
  • Compatible WithHoneywell dual-input controller
  • TypeOutput—two relays & 0/4 to 20 mA
  • DescriptionTwo Relays & 0/4 to 20 mA Output Card,