Hoffman CONCEPT Steel Wall-Mount Enclosures, Types 4, 12
Ideal for machine control applications
  • Available in 14, 16 or 18 gauge steel
  • Seams continuously welded and ground smooth
  • Simple easy-to-remove and install hinge pins with built-in captivation clip
  • High-torque thread-less studs and fasteners on door
  • Minimum-width external formed body flange provides maximum door opening (210 degrees)
  • Panel mounting studs fit optional CONCEPT panels and other accessories; mounting holes in back of body for optional external wall-mount brackets
  • Seamless foam-in-place gasket
  • Solid corner-formed doors are interchangeable and easily removed by pulling clipstyle hinge pins
  • Quarter-turn slotted latch(es)
  • Grounding stud on body; bonding provision on door
  • Provisions for thermoplastic data pocket
  • Ratings: NEMA type 4, 12; IP66
These mild steel enclosures feature streamlined styling, flush quarter-turn latches, and an attractive and durable RAL 7035 textured light-gray polyester powder paint finish inside and out.
122,000.00 - ₹146,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Depth (in) Outside Height (in) Outside Width (in)
Availability Pricing
nVent Hoffman CSD20168LG Concept Steel NEMA 4/12 Enclosure, 1/4-Turn, 20x16x8" WW-10500-48 Mfr # CSD20168LG
COMPARE
8 20 16
₹122,000.00INR / Each
nVent Hoffman CSD242010LG Concept Steel NEMA 4/12 Enclosure, 1/4-Turn, 24x20x10" WW-10502-40 Mfr # CSD242010LG
COMPARE
10 24 20
₹146,000.00INR / Each