nVent Hoffman CSD20168LG Concept Steel NEMA 4/12 Enclosure, 1/4-Turn, 20x16x8"

Representative image only.

nVent Hoffman CSD20168LG Concept Steel NEMA 4/12 Enclosure, 1/4-Turn, 20x16x8"

nVent Hoffman | Mfr: CSD20168LG| Item: 1050048

Ideal for machine control applications

₹167,000.00INR / Each

₹167,000.00INR / Each
Product Description

Includes

hardware kit with panel mounting nuts, panel grounding hardware, sealing washers, and illustrated instruction sheet.

Key Features

  • Available in 14, 16 or 18 gauge steel
  • Seams continuously welded and ground smooth
  • Simple easy-to-remove and install hinge pins with built-in captivation clip
  • High-torque thread-less studs and fasteners on door
  • Minimum-width external formed body flange provides maximum door opening (210 degrees)
  • Panel mounting studs fit optional CONCEPT panels and other accessories; mounting holes in back of body for optional external wall-mount brackets
  • Seamless foam-in-place gasket
  • Solid corner-formed doors are interchangeable and easily removed by pulling clipstyle hinge pins
  • Quarter-turn slotted latch(es)
  • Grounding stud on body; bonding provision on door
  • Provisions for thermoplastic data pocket
  • Ratings: NEMA type 4, 12; IP66
  • 1 year warranty

More About this Item

These mild steel enclosures feature streamlined styling, flush quarter-turn latches, and an attractive and durable RAL 7035 textured light-gray polyester powder paint finish inside and out.

Note: Optional mounting brackets are required to meet UL and CSA external mounting requirements;

Conformite Europeenne
Specifications
  • MaterialSteel
  • Door Gauge (Gauge)16
  • Outside Width (in)16
  • Outside Height (in)20
  • Outside Depth (in)8
  • DescriptionConcept Steel NEMA 4/12 Enclosure, 1/4-Turn, 20x16x8"
  • Warranty1 Year
Reviews
Q&A