High-Tech Conversions Lint-Free Nonwoven Cleanroom Wipes
Durable and highly absorbent—suitable for a variety of cleanroom applications
  • Cleanroom ISO Class 5 compliant Polyester and cellulose blend provides strong fabric free of particle generation
Use these lint-free nonwoven cleanroom wipes for general-purpose cleaning applications in ISO Class 5 cleanrooms or higher. The synergistic blend of virgin polyester and cellulose is hydro-entangled to form a super-tough fabric that is highly absorbent and virtually free of particle generation. The ISO Class 5 certified fabric is also cleanroom laundered and abrasion resistant.
72,300.00 - ₹88,300.00INR / Case of 14400
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (in) Material Width (in)
Availability Pricing
High-Tech Conversions NT1-44 Cleanroom wipes, non-woven, lint-free, polyester/ cellulose blend, 4" x 4", 1200 wipes per bag, 14400/CS 1598124 Mfr # NT1-44
COMPARE
4 Polyester/Cellulose 4
₹88,300.00INR / Case of 14400
High-Tech Conversions NT1-66 Cleanroom wipes, non-woven, lint-free, polyester/ cellulose blend, 6" x 6", 600 wipes per bag, 6000/CS 1598125 Mfr # NT1-66
COMPARE
6 Polyester/Cellulose 6
₹74,800.00INR / Case of 6000
High-Tech Conversions NT1-99 Cleanroom wipes, non-woven, lint-free, polyester/ cellulose blend, 9" x 9", 300 wipes per bag, 3600/CS 1598126 Mfr # NT1-99
COMPARE
9 Polyester/Cellulose 9
₹72,300.00INR / Case of 3600
High-Tech Conversions NT10-1212 Cleanroom wipes, non-woven, lint-free, polyester/ cellulose blend, 12" x 12", 150 wipes per bag, 2100/CS 1598127 Mfr # NT1-1212
COMPARE
12 Polyester/Cellulose 12
₹82,800.00INR / Case of 2100
High-Tech Conversions NT1-1818 Cleanroom wipes, non-woven, lint-free, polyester/ cellulose blend, 18" x 18", 150 wipes per bag, 750/CS 1598128 Mfr # NT1-1818
COMPARE
18 Polyester/Cellulose 18
₹74,800.00INR / Case of 750