High-Tech Conversions EDS Cleanroom Paper

Ideal of use in electronics assembly cleanrooms
  • IPA resistance
  • Compatible to most inks in writing without smearing
  • Ultra-low particle and fiber generation
  • Compatible with laser and inkjet printers
  • Class 100 (ISO 5) compliant
  • Latex-free
The EDS safe cleanroom paper offers ultra-low ionic contamination and features excellent heat resistance material for laser printing and photocopiers. It is compatible with most pens as well as inkjet printers. Double bagged, vacuum packed, and processed in a Class 100 (ISO 5) controlled environment, the paper is suitable for use in a variety of electronics assembly cleanrooms. The 22 weight paper is available in blue, white, and green.
42,800.00INR / Case of 2500

3 variations of this product are available.

See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Color Length (in) Width (in)
Availability Pricing
High-Tech Conversions Bee-Write™ EDS Cleanroon Paper, 22 Weight, 8.5" x 11", Blue; 2500/CS 3367870 Mfr # BW-CRP-8511-BLUE
COMPARE
Blue 11 8.5
₹42,800.00INR / Case of 2500
High-Tech Conversions Bee-Write™ EDS Cleanroon Paper, 22 Weight, 8.5" x 11", White; 2500/CS 3367871 Mfr # BW-CRP-8511-WHITE
COMPARE
White 11 8.5
₹42,800.00INR / Case of 2500
High-Tech Conversions Bee-Write™ EDS Cleanroon Paper, 22 Weight, 8.5" x 11", Green; 2500/CS 3367872 Mfr # BW-CRP-8511-GREEN
COMPARE
Green 11 8.5
₹42,800.00INR / Case of 2500
Showing 1- 3 of 3

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat