Hamilton® SoftGrip™ Fixed-Volume Pipettors
Contoured,ergonomical design provides comfortable, fatigue-free operation
  • Color coding allows easy volume identification
  • Autoclave entire pipettor for total decontamination and sterilization
SoftGrip™ pipettors assure you of quality, comfort and precision manual pipetting. The slim ejection sleeve is made from a high strength, chemically resistant polymer. The slim design allows the pipette to reach to the bottom of narrow tubes without the need for extended length tips. SoftGrip pipettes have a universal calibration key that reduces the need for costly calibration services. Calibration requires only minutes to perform and eliminates the downtime and cost of alternative calibration methods. The SoftGrip tip nozzle ensures a proper fit with universal pipette tips. Unique color coding and labeling clearly indicates the volume of the pipettes.
32,800.00INR / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy (%) Color Code Max Volume (µL)
Availability Pricing
Hamilton SoftGrip™ Fixed-Volume Pipettor, Sandstone, 50 µL WW-25704-33 Mfr # 55019-07
COMPARE
±0.60% Sandstone 50
₹32,800.00INR / Each
Hamilton SoftGrip™ Fixed-Volume Pipettor, Violet, 100 µL WW-25704-34 Mfr # 55019-09
COMPARE
±0.50% Violet 100
₹32,800.00INR / Each
Hamilton SoftGrip™ Fixed-Volume Pipettor, Brick Red, 300 µL WW-25704-37 Mfr # 55019-15
COMPARE
±0.40% Brick Red 300
₹32,800.00INR / Each
Hamilton SoftGrip™ Fixed-Volume Pipettor, Olive Green, 500 µL WW-25704-38 Mfr # 55019-17
COMPARE
±0.40% Olive Green 500
₹32,800.00INR / Each
Hamilton SoftGrip™ Fixed-Volume Pipettor, Sky Blue, 1000 µL WW-25704-39 Mfr # 55019-19
COMPARE
±0.30% Sky Blue 1000
₹32,800.00INR / Each